4-Chloro-1H-pyrazole ≥98.0%

Leverantör: TCI EUROPE
Varning

C2888-5G C2888-25G
TCIAC2888-5GEA 765 SEK
TCIAC2888-5G TCIAC2888-25G
4-Chloro-1H-pyrazole ≥98.0%
4-Chloro-1H-pyrazole
Formel: C₃H₃ClN₂
MW: 102,5200 g/mol
Kokpunkt: 220°C
MDL Number: MFCD00159634
CAS nummer: 15878-00-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR