Kloretan ∼15% ∼2 mol/L i THF
Leverantör: TCI
|
Fara
|
Synonyms:
Etylklorid
C2883-100ML
C2883-500ML
TCIAC2883-100MLEA
1100
SEK
TCIAC2883-100ML
TCIAC2883-500ML
Kloretan ∼15% ∼2 mol/L i THF
Kloretan
Formel:
C₂H₅Cl Storage Temperature: Kylskåp |
MDL Number:
MFCD00000961 CAS nummer: 75-00-3 EINECS: 200-830-5 UN: 1993 ADR: 3,II Merck Index: 14,03782 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Concentration (HNMR) | 14.5 to 15.5 w/w% |
Water | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...