Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Benzyl 3-pyrroline-1-carboxylate, 1H-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester, benzyl 2,5-dihydropyrrole-1-carboxylate, Benzyl 2,5-dihydro-1H-pyrrole-1-carboxylate
C2872-1G C2872-5G
TCIAC2872-1GEA 790 SEK
TCIAC2872-1G TCIAC2872-5G
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥97.0%
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Formel: C₁₂H₁₃NO₂
MW: 203,241 g/mol
Kokpunkt: 126°C
MDL Number: MFCD01863559
CAS nummer: 31970-04-4
Order Now
Specifikation testresultat
Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %