λ-Carrageenan, low viscosity
Leverantör: TCI
Synonyms:
lambda-Carrageenan (High-viscosity)
C2871-1G
C2871-5G
TCIAC2871-1GEA
495
SEK
TCIAC2871-1G
TCIAC2871-5G
λ-Carrageenan, low viscosity
lambda-Carrageenan
MDL Number:
MFCD00151513 CAS nummer: 9064-57-7 EINECS: 232-953-5 Merck Index: 14,01864 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Viscosity | min. 10 mPa-s (0.3 %, H2O, 25 deg-C) |
Drying loss | max. 15.0 % |
Ash Content | max. 30.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...