Clofazimine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine, N,5-Bis(4-chlorophenyl)-3-(isopropylimino)-3,5-dihydrophenazin-2-amine
C2866-1G
C2866-5G
TCIAC2866-1GEA
830
SEK
TCIAC2866-1G
TCIAC2866-5G
Clofazimine ≥98.0% (by HPLC, titration analysis)
Clofazimine
Formel:
C₂₇H₂₂Cl₂N₄ MW: 473,40 g/mol |
MDL Number:
MFCD00056793 CAS nummer: 2030-63-9 EINECS: 217-980-2 Merck Index: 14,02373 |
Specifikation testresultat
Appearance | Yellow to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...