1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%, purified by sublimation

Leverantör: TCI EUROPE
Varning

Synonyms: Cyclobuta[1,2-c:3,4-c']difuran-1,3,4,6(3aH,3bH,6aH,6bH)-tetraone

C2842-1G C2842-5G
TCIAC2842-1GEA 655 SEK
TCIAC2842-1G TCIAC2842-5G
1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%, purified by sublimation
1,2,3,4-Cyclobutanetetracarboxylic dianhydride
Formel: C₈H₄O₆
MW: 196,11 g/mol
Smältpunkt: 285°C
MDL Number: MFCD00004944
CAS nummer: 4415-87-6
EINECS: 224-577-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53eeeb976d6709496950f801c2dc6f2721eee18c&vid=c77e6554cb19ced5c0b2ca8126f249ec64c92f01&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR