N-Carbobenzoxyglycine methyl ester ≥95.0%

Leverantör: TCI EUROPE

Synonyms: N-Cbz-glycine Methyl Ester, N-Benzyloxycarbonylglycine methyl ester, Z-Gly-OMe

C2838-5G C2838-25G
TCIAC2838-5GEA 335 SEK
TCIAC2838-5G TCIAC2838-25G
N-Carbobenzoxyglycine methyl ester ≥95.0%
N-Carbobenzoxyglycine methyl ester
Formel: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Kokpunkt: 151°C
MDL Number: MFCD00043631
CAS nummer: 1212-53-9
EINECS: 214-922-8

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR