4-Chloro-2-methylpyrimidine ≥98.0%
Leverantör: TCI
Varning
C2835-200MG C2835-1G
TCIAC2835-200MGEA 310 SEK
TCIAC2835-200MG TCIAC2835-1G
4-Chloro-2-methylpyrimidine ≥98.0%
4-Chloro-2-methylpyrimidine
Formel: C₅H₅ClN₂
MW: 128.5596 g/mol
Kokpunkt: 168°C
MDL Number: MFCD00234069
CAS nummer: 4994-86-9
Order Now
Specifikation testresultat
Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C