3-Cyanoumbelliferone 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 3-Cyano-7-hydroxycoumarin, 7-Hydroxy-2-oxo-2H-chromene-3-carbonitrile

C2834-1G C2834-5G
TCIAC2834-1GEA 1475 SEK
TCIAC2834-1G TCIAC2834-5G
3-Cyanoumbelliferone 98,0 GC_ANALYSMETOD
3-Cyanoumbelliferone
Formel: C₁₀H₅NO₃
MW: 187.15 g/mol
MDL Number: MFCD00037480
CAS nummer: 19088-73-4

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR