3-Cyanocoumarin ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Coumarin-3-carbonitrile, 2-Oxo-2H-chromene-3-carbonitrile
C2830-1G
C2830-5G
TCIAC2830-1GEA
285
SEK
TCIAC2830-1G
TCIAC2830-5G
3-Cyanocoumarin ≥98.0%
3-Cyanocoumarin
Formel:
C₁₀H₅NO₂ MW: 171,16 g/mol |
MDL Number:
MFCD00115699 CAS nummer: 15119-34-3 |
Specifikation testresultat
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...