2-Cyano-N,N-diethylacetamide ≥98.0%

Leverantör: TCI
Varning

C2822-5G C2822-25G
TCIAC2822-5GEA 510 SEK
TCIAC2822-5G TCIAC2822-25G
2-Cyano-N,N-diethylacetamide ≥98.0%
2-Cyano-N,N-diethylacetamide
Formel: C₇H₁₂N₂O
Kokpunkt: 114°C
Storage Temperature: Kylskåp
MDL Number: MFCD01332383
CAS nummer: 26391-06-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR