Cyclohexadiene (1,3- and 1,4- mixture) ≥85.0% (contains 4-10% Benzene) stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: Dihydrobenzene

Stabilization: Stabilised med BHT/Ionol

C2808-5ML C2808-25ML
TCIAC2808-5MLEA 1025 SEK
TCIAC2808-5ML TCIAC2808-25ML
Cyclohexadiene (1,3- and 1,4- mixture) ≥85.0% (contains 4-10% Benzene) stabilised
Cyclohexadiene
Formel: C₆H₈
MW: 80,13 g/mol
Storage Temperature: Kylskåp
UN: 3295
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 % (total of isomer)
Benzene 4.0 to 10.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR