4-Cyano-4'-propylbiphenyl ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4'-Propyl-[1,1'-biphenyl]-4-carbonitrile

C2764-1G C2764-5G
TCIAC2764-1GEA 430 SEK
TCIAC2764-1G TCIAC2764-5G
4-Cyano-4'-propylbiphenyl ≥99.0%
4-Cyano-4'-propylbiphenyl
Formel: C₁₆H₁₅N
MW: 221,2970 g/mol
MDL Number: MFCD00505987
CAS nummer: 58743-76-3
EINECS: 261-415-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 65.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR