(2-Chlorophenyl)acetylene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Chloro-2-ethynylbenzene
C2750-1G C2750-5G
TCIAC2750-1GEA 375 SEK
TCIAC2750-1G TCIAC2750-5G
(2-Chlorophenyl)acetylene ≥98.0%
(2-Chlorophenyl)acetylene
Formel: C₈H₅Cl
MW: 136,58 g/mol
Kokpunkt: 72°C
Flampunkt: 70°C
Storage Temperature: Kylskåp
MDL Number: MFCD00269951
CAS nummer: 873-31-4
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %