5-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Chloro-2-pyridinecarboxylic acid, 5-Chloropyridine-2-carboxylic acid, 5-Chloro-2-picolinic acid
C2736-1G
C2736-5G
TCIAC2736-1GEA
245
SEK
TCIAC2736-1G
TCIAC2736-5G
5-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
5-Chloropicolinic acid
Formel:
C₆H₄ClNO₂ MW: 157,56 g/mol |
MDL Number:
MFCD04114192 CAS nummer: 86873-60-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 171.0 to 175.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...