1-Chloro-4-iodobutane ≥98.0% stabilised
Leverantör: TCI
Varning
Stabilization: Stabilised with copper chip
C2713-5G C2713-25G
TCIAC2713-5GEA 470 SEK
TCIAC2713-5G TCIAC2713-25G
1-Chloro-4-iodobutane ≥98.0% stabilised
1-Chloro-4-iodobutane
Formel: C₄H₈ClI
MW: 218,46 g/mol
Kokpunkt: 67°C
Flampunkt: 93°C
MDL Number: MFCD00039415
CAS nummer: 10297-05-9
EINECS: 233-669-4
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %