6-(Trifluormethyl)nicotinonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
5-Cyano-2-(trifluoromethyl)pyridine, 6-(Trifluoromethyl)nicotinonitrile
C2698-1G
C2698-5G
TCIAC2698-1GEA
830
SEK
TCIAC2698-1G
TCIAC2698-5G
6-(Trifluormethyl)nicotinonitrile ≥98.0%
6-(Trifluormethyl)nicotinonitrile
Formel:
C₇H₃F₃N₂ MW: 172,1100 g/mol |
MDL Number:
MFCD03001347 CAS nummer: 216431-85-5 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...