2-Chloro-5-methoxypyrimidine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C2693-1G
C2693-5G
TCIAC2693-1GEA
565
SEK
TCIAC2693-1G
TCIAC2693-5G
2-Chloro-5-methoxypyrimidine ≥98.0%
2-Chloro-5-methoxypyrimidine
Formel:
C₅H₅ClN₂O MW: 144,56 g/mol |
MDL Number:
MFCD08702770 CAS nummer: 22536-65-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...