3-Cyano-4-fluorobenzaldehyde ≥98.0%
Leverantör: TCI
|
Varning
|
C2692-1G
C2692-5G
TCIAC2692-1GEA
350
SEK
TCIAC2692-1G
TCIAC2692-5G
3-Cyano-4-fluorobenzaldehyde ≥98.0%
3-Cyano-4-fluorobenzaldehyde
Formel:
C₈H₄FNO |
MDL Number:
MFCD01863558 CAS nummer: 218301-22-5 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...