5-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: (5-Chloro-2-methylphenyl)boronic acid, 5-Chloro-2-methylbenzeneboronic acid

C2665-1G C2665-5G
TCIAC2665-1GEA 430 SEK
TCIAC2665-1G TCIAC2665-5G
5-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-methylphenylboronic acid
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
Smältpunkt: 186°C
MDL Number: MFCD03411939
CAS nummer: 148839-33-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR