3-Cyano-6-isopropylchromone ≥98.0%

Leverantör: TCI
Varning

C2661-1G C2661-5G
TCIAC2661-1GEA 685 SEK
TCIAC2661-1G TCIAC2661-5G
3-Cyano-6-isopropylchromone ≥98.0%
3-Cyano-6-isopropylchromone
Formel: C₁₃H₁₁NO₂
MDL Number: MFCD00191961
CAS nummer: 50743-32-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR