3-Cyclohexyl-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: D-CHA-OH Hydrate, (R)-2-Amino-3-cyclohexylpropanoic acid hydrate(1:x)

C2659-1G C2659-5G
TCIAC2659-1GEA 750 SEK
TCIAC2659-1G TCIAC2659-5G
3-Cyclohexyl-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
3-Cyclohexyl-D-alanine hydrate
Formel: C₉H₁₉NO₃
MW: 189,25 g/mol
MDL Number: MFCD00217056
CAS nummer: 213178-94-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -11.0 to -13.0 deg (C=1、1mol/L HCl) (calcd.on anh.substance)
Water 3.0 to 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR