Cyclopentylmethanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Cyclopentanemethanol
C2658-5G
C2658-25G
TCIAC2658-5GEA
615
SEK
TCIAC2658-5G
TCIAC2658-25G
Cyclopentylmethanol ≥98.0%
Cyclopentylmethanol
Formel:
C₆H₁₂O MW: 100,16 g/mol Kokpunkt: 163°C Flampunkt: 62°C |
MDL Number:
MFCD00001384 CAS nummer: 3637-61-4 EINECS: 222-861-3 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...