4-Chlorosalicylaldehyde ≥95.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
4-Chloro-2-hydroxybenzaldehyde
C2653-1G
C2653-5G
TCIAC2653-1GEA
605
SEK
TCIAC2653-1G
TCIAC2653-5G
4-Chlorosalicylaldehyde ≥95.0% (by GC, titration analysis)
4-Chlorosalicylaldehyde
Formel:
C₇H₅ClO₂ |
MDL Number:
MFCD06252499 CAS nummer: 2420-26-0 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...