Candesartan cilexetil ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-(((Cyclohexyloxy)carbonyl)oxy)ethyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate
C2635-200MG
C2635-1G
TCIAC2635-200MGEA
485
SEK
TCIAC2635-200MG
TCIAC2635-1G
Candesartan cilexetil ≥98.0% (by HPLC, titration analysis)
Candesartan cilexetil
Formel:
C₃₃H₃₄N₆O₆ MW: 610,67 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00871371 CAS nummer: 145040-37-5 Merck Index: 14,01739 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 163.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...