4-Cyano-4'-octyloxybiphenyl ≥98.0%
Leverantör: TCI
|
Varning
|
C2618-5G
C2618-25G
TCIAC2618-5GEA
895
SEK
TCIAC2618-5G
TCIAC2618-25G
4-Cyano-4'-octyloxybiphenyl ≥98.0%
4-Cyano-4'-octyloxybiphenyl
Formel:
C₂₁H₂₅NO Smältpunkt: 54°C |
MDL Number:
MFCD00075145 CAS nummer: 52364-73-5 EINECS: 257-878-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...