2-Chloro-6-fluorobenzothiazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Chloro-6-fluorobenzo[d]thiazole
C2607-1G
C2607-5G
TCIAC2607-1GEA
950
SEK
TCIAC2607-1G
TCIAC2607-5G
2-Chloro-6-fluorobenzothiazole ≥98.0%
2-Chloro-6-fluorobenzothiazole
Formel:
C₇H₃ClFNS MW: 187,62 g/mol |
MDL Number:
MFCD04448822 CAS nummer: 399-74-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...