Cilostazol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 6-(4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

C2587-5G C2587-1G
TCIAC2587-5GEA 1850 SEK
TCIAC2587-5G TCIAC2587-1G
Cilostazol ≥98.0%
Cilostazol
Formel: C₂₀H₂₇N₅O₂
MW: 369,47 g/mol
MDL Number: MFCD00866780
CAS nummer: 73963-72-1
Merck Index: 14,02277

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR