4-Chlorobenzoylacetonitrile ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4-Chlorophenacylcyanide, 3-(4-Chlorophenyl)-3-oxopropanenitrile
C2582-5G C2582-25G
TCIAC2582-5GEA 3950 SEK
TCIAC2582-5G TCIAC2582-25G
4-Chlorobenzoylacetonitrile ≥98.0%
4-Chlorobenzoylacetonitrile
Formel: C₉H₆ClNO
MW: 179,61 g/mol
Smältpunkt: 128°C
Storage Temperature: Kylskåp
MDL Number: MFCD00051625
CAS nummer: 4640-66-8
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %