Carazolol ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-(9H-Carbazol-4-yloxy)-3-(isopropylamino)-2-propanol

C2578-200MG C2578-1G
TCIAC2578-200MGEA 1325 SEK
TCIAC2578-200MG TCIAC2578-1G
Carazolol ≥98.0% (by HPLC, titration analysis)
Carazolol
Formel: C₁₈H₂₂N₂O₂
MW: 298,39 g/mol
MDL Number: MFCD00864588
CAS nummer: 57775-29-8
EINECS: 260-945-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,01778

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR