4-Carbamoylphenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI
|
Varning
|
Synonyms:
(4-carbamoylphenyl)boronic Acid, 4-Aminocarbonylphenylboronic acid
C2570-1G
C2570-5G
TCIAC2570-1GEA
605
SEK
TCIAC2570-1G
TCIAC2570-5G
4-Carbamoylphenylboronic acid (contains varying amounts of Anhydride)
4-Carbamoylphenylboronic acid
Formel:
C₇H₈BNO₃ Smältpunkt: 234°C Storage Temperature: Kylskåp |
MDL Number:
MFCD03411940 CAS nummer: 123088-59-5 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...