Chloridazon ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Amino-4-chloro-2-phenylpyridazin-3(2H)-one, Pyrazon

C2554-1G C2554-5G
TCIAC2554-1GEA 870 SEK
TCIAC2554-1G TCIAC2554-5G
Chloridazon ≥98.0%
Chloridazon
Formel: C₁₀H₈ClN₃O
MW: 221,65 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD02668021
CAS nummer: 1698-60-8
EINECS: 216-920-2
UN: 3077
ADR: 9,III

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 210.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR