8-Aminocaprylic acid 99+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid
226830010 226830050
ACRO226830010EA 840 SEK
ACRO226830010 ACRO226830050
8-Aminocaprylic acid 99+%
8-Aminocaprylic acid
Formel: C₈H₁₇NO₂
MW: 159.23 g/mol
Kokpunkt: 289
Smältpunkt: 194…196 °C
Densitet: 1.221
MDL Number: MFCD00008245
CAS nummer: 1002-57-9
EINECS: 213-687-9
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %