6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one 95,0 HPLC, N

Leverantör: TCI
Varning

C2404-200MG C2404-1G
TCIAC2404-200MGEA 1850 SEK
TCIAC2404-200MG TCIAC2404-1G
6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one 95,0 HPLC, N
6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one
Formel: C₂₁H₁₇N₃O₂
MW: 343.39 g/mol
CAS nummer: 128119-95-9

Order Now

Specifikation testresultat

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR