2-Chlorophenyl cyclopentyl ketone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Chlorobenzoylcyclopentane

C2338-5G
TCIAC2338-5GEA 2400 SEK
TCIAC2338-5G
2-Chlorophenyl cyclopentyl ketone ≥98.0%
2-Chlorophenyl cyclopentyl ketone
Formel: C₁₂H₁₃ClO
MW: 208,69 g/mol
Kokpunkt: 96°C
MDL Number: MFCD00038367
CAS nummer: 6740-85-8
EINECS: 229-802-0

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR