6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine
C2306-1G
C2306-5G
TCIAC2306-1GEA
735
SEK
TCIAC2306-1G
TCIAC2306-5G
6-Chloro-7-deazapurine ≥98.0% (by GC, titration analysis)
6-Chloro-7-deazapurine
Formel:
C₆H₄ClN₃ MW: 153,57 g/mol Smältpunkt: 188°C |
MDL Number:
MFCD01686865 CAS nummer: 3680-69-1 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...