4-Chloroquinoline ≥97.0%

Leverantör: TCI EUROPE
Varning

C2284-1G C2284-5G
TCIAC2284-1GEA 575 SEK
TCIAC2284-1G TCIAC2284-5G
4-Chloroquinoline ≥97.0%
4-Chloroquinoline
Formel: C₉H₆ClN
MW: 163,61 g/mol
Kokpunkt: 261°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006773
CAS nummer: 611-35-8
EINECS: 210-267-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df5ba2139acca291b0f9d1224c81e7ecf9e2fd8a&vid=df5ba2139acca291b0f9d1224c81e7ecf9e2fd8a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR