2-Chloro-N-methoxy-N-methylacetamide ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 2-Chloro-N-methylacetohydroxamate
C2272-25G C2272-5G
TCIAC2272-25GEA 1875 SEK
TCIAC2272-25G TCIAC2272-5G
2-Chloro-N-methoxy-N-methylacetamide ≥98.0%
2-Chloro-N-methoxy-N-methylacetamide
Formel: C₄H₈ClNO₂
MW: 137,56 g/mol
Kokpunkt: 95°C
Storage Temperature: Kylskåp
MDL Number: MFCD00134232
CAS nummer: 67442-07-3
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C