1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Cyclobuta[1,2-c:3,4-c']difuran-1,3,4,6(3aH,3bH,6aH,6bH)-tetraone
C2262-1G
C2262-5G
TCIAC2262-1GEA
455
SEK
TCIAC2262-1G
TCIAC2262-5G
1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0%
1,2,3,4-Cyclobutanetetracarboxylic dianhydride
Formel:
C₈H₄O₆ MW: 196,11 g/mol Smältpunkt: 285°C |
MDL Number:
MFCD00004944 CAS nummer: 4415-87-6 EINECS: 224-577-5 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Anilide Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...