Clindamycin hydrochloride monohydrate ≥98.0%

Leverantör: TCI EUROPE

C2256-25G C2256-5G
TCIAC2256-25GEA 2825 SEK
TCIAC2256-25G TCIAC2256-5G
Clindamycin hydrochloride monohydrate ≥98.0%
Clindamycin hydrochloride monohydrate
Formel: C₁₈H₃₃ClN₂O₅S
MW: 461,44 g/mol
Smältpunkt: 143°C
MDL Number: MFCD06411091
CAS nummer: 58207-19-5
Merck Index: 14,02356

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +140 to +147 deg (C=2, H2O) (calcd.on anh.substance)
Water 3.0 to 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR