Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)

Leverantör: TCI EUROPE
Varning

C2251-1G C2251-250MG
TCIAC2251-1GEA 1900 SEK
TCIAC2251-1G TCIAC2251-250MG
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Formel: C₅₅H₄₇OP₃Ru
MW: 917,97 g/mol
MDL Number: MFCD00015870
CAS nummer: 25360-32-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Elemental analysis (Carbon) 69.8 to 73.4 %
Functionality test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR