Gimeracil ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 5-Chloro-4-hydroxypyridin-2(1H)-one, 5-Chloro-4-hydroxy-2(1H)-pyridinone, 5-Chloro-2,4-pyridinediol, 5-Chloro-2,4-dihydroxypyridine

C2243-1G C2243-5G
TCIAC2243-1GEA 950 SEK
TCIAC2243-1G TCIAC2243-5G
Gimeracil ≥98.0% (by HPLC, titration analysis)
Gimeracil
Formel: C₅H₄ClNO₂
MW: 145,54 g/mol
Smältpunkt: 274°C
MDL Number: MFCD08458352
CAS nummer: 103766-25-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR