Carbon nanotube multi-walled, Length: 5-15 µm, Ø 20-40 nm
Leverantör: TCI
Varning
Synonyms: CNT
C2154-5G C2154-1G
TCIAC2154-5GEA 2750 SEK
TCIAC2154-5G TCIAC2154-1G
Carbon nanotube multi-walled, Length: 5-15 µm, Ø 20-40 nm
Carbon nanotube
Formel: C
MW: 12.011 g/mol
MDL Number: MFCD00133992
CAS nummer: 308068-56-6
Order Now
Specifikation testresultat
Appearance Black powder to crystal
Purity (TPO Method) min. 95 %
Diameter 20 to 40 nm
Length 5 to 15 micro_m