N-Carbobenzoxy-D-threonine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
Z-D-Thr-OH, N-Cbz-D-threonine, (2R,3S)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxybutanoic acid
C2138-1G
C2138-5G
TCIAC2138-1GEA
285
SEK
TCIAC2138-1G
TCIAC2138-5G
N-Carbobenzoxy-D-threonine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-D-threonine
Formel:
C₁₂H₁₅NO₅ MW: 253,25 g/mol |
MDL Number:
MFCD00037808 CAS nummer: 80384-27-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Specific rotation [a]20/D | +4.0 to +7.0 deg (C=2, AcOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...