3-Chloro-2-hydroxypropyl methacrylate ≥83.0% stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: Methacrylic Acid 3-Chloro-2-hydroxypropyl Ester

Stabilization: med hydrokinon

C2105-25G C2105-500G
TCIAC2105-25GEA 215 SEK
TCIAC2105-25G TCIAC2105-500G
3-Chloro-2-hydroxypropyl methacrylate ≥83.0% stabilised
3-Chloro-2-hydroxypropyl methacrylate
Formel: C₇H₁₁ClO₃
MW: 178,61 g/mol
Kokpunkt: 108°C
Flampunkt: 138°C
Storage Temperature: Kylskåp
MDL Number: MFCD00040686
CAS nummer: 13159-52-9
EINECS: 236-097-3

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 83.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR