Carboplatin ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)
C2043-1G
C2043-100MG
TCIAC2043-1GEA
4250
SEK
TCIAC2043-1G
TCIAC2043-100MG
Carboplatin ≥98.0%
Carboplatin
Formel:
C₆H₁₂N₂O₄Pt MW: 371,26 g/mol |
MDL Number:
MFCD00070464 CAS nummer: 41575-94-4 EINECS: 255-446-0 Merck Index: 14,01822 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...