2-(4-Chlorophenyl)benzimidazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
2-(4-Chlorophenyl)-1H-benzo[d]imidazole
C2027-1G
C2027-5G
TCIAC2027-1GEA
1750
SEK
TCIAC2027-1G
TCIAC2027-5G
2-(4-Chlorophenyl)benzimidazole ≥98.0% (by GC, titration analysis)
2-(4-Chlorophenyl)benzimidazole
Formel:
C₁₃H₉ClN₂ MW: 228,68 g/mol Smältpunkt: 303°C |
MDL Number:
MFCD00224359 CAS nummer: 1019-85-8 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...