Iota carrageenan

Leverantör: TCI

C1805-25G C1805-500G
TCIAC1805-25GEA 215 SEK
TCIAC1805-25G TCIAC1805-500G
Iota carrageenan
Iota carrageenan
MDL Number: MFCD00151512
CAS nummer: 9062-07-1
EINECS: 232-949-3
Merck Index: 14,01858

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Drying loss max. 12.0 %
Viscosity 5.0 to 20.0 m Pa-s (0.3 %, H2O, 25 deg-C)
Ash Content max. 40.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR