Ethyl-(S)-4-Chloro-3-hydroxybutyrate ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(S)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester, Ethyl (S)-4-chloro-3-hydroxybutanoate
C1717-25G
C1717-5G
TCIAC1717-25GEA
1050
SEK
TCIAC1717-25G
TCIAC1717-5G
Ethyl-(S)-4-Chloro-3-hydroxybutyrate ≥95.0%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate
Formel:
C₆H₁₁ClO₃ MW: 166,60 g/mol Kokpunkt: 96°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00211241 CAS nummer: 86728-85-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Specific rotation [a]20/D | -13.0 to -16.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...