4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥98.0%

Leverantör: TCI
Fara

Synonyms: 1-Cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-Tetrahydrophthalic anhydride

C1701-5G C1701-25G
TCIAC1701-5GEA 230 SEK
TCIAC1701-5G TCIAC1701-25G
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥98.0%
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione
Formel: C₈H₈O₃
MW: 152,15 g/mol
MDL Number: MFCD00005917
CAS nummer: 2426-02-0
EINECS: 219-374-3
UN: 2698
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 96.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR